Assay Details
Target Gene Details
Entrez Gene ID: | 67819 |
Gene Name: | Der1-like domain family, member 1 |
Gene Aliases: |
1110021N07Rik, AI195141, AW551338, Derlin-1 |
Location: |
Chr.15:57869502-57892418 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Derl1 | NM_024207.4 | 1 | 48 | NP_077169.1 |
| AK007435.1 | 1 | 53 | BAB25036.1 | |
| AK011499.1 | 1 | 70 | ||
| AK043420.1 | 1 | 49 | BAC31545.1 | |
| AK075631.1 | BAC35868.1 | |||
| AK148018.1 | BAE28291.1 | |||
| AK152197.1 | 1 | 46 | BAE31026.1 | |
| AK152901.1 | BAE31582.1 | |||
| AK161702.1 | 1 | 74 | ||
| AK219480.1 | ||||
| BC003454.1 | 1 | 73 | AAH03454.1 | |
| BC085490.1 | 1 | 78 | AAH85490.1 |
More Information
Set Membership: |
|

Genomic Map