Assay Details
Target Gene Details
Entrez Gene ID: | 554292 |
Gene Name: | methyltransferase hypoxia inducible domain containing 1 |
Gene Aliases: |
Mettl7a2-Higd1c, Mettl7a2Higd1c, Mettl7a2higd1c, UbiE-YGHL1 |
Location: |
Chr.15:100353200-100384435 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Methig1 | NM_001024672.3 | 4 | 1093 | NP_001019843.1 |
| AK144032.1 | 4 | 1094 | BAE25664.1 |
More Information
Set Membership: |
|

Genomic Map