Assay Details
Target Gene Details
Entrez Gene ID: | 23970 |
Gene Name: | protein kinase C and casein kinase substrate in neurons 2 |
Gene Aliases: |
AI197433 |
Location: |
Chr.15:83375607-83464606 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pacsin2 | NM_001159509.1 | 11 | 2277 | NP_001152981.1 |
| NM_001159510.1 | 11 | 2228 | NP_001152982.1 | |
| NM_011862.3 | 11 | 2316 | NP_035992.1 | |
| AF128535.1 | 11 | 2306 | AAD41780.1 | |
| AK083414.1 | 3 | 966 | BAC38907.1 | |
| AK154785.1 | 11 | 2203 | BAE32828.1 | |
| AK170299.1 | BAE41697.1 | |||
| BC023502.1 | 12 | 2182 | AAH23502.1 |
More Information
Set Membership: |
|

Genomic Map