Assay Details
Target Gene Details
Entrez Gene ID: | 56722 |
Gene Name: | LPS-induced TN factor |
Gene Aliases: |
3222402J11Rik, C85531, N4WBP3, TBX1 |
Location: |
Chr.16:10959273-10993121 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Litaf | NM_019980.2 | 4 | 1644 | NP_064364.1 |
| AF171100.1 | 4 | 1612 | AAF27312.1 | |
| AK018578.1 | 4 | 1627 | BAB31289.1 | |
| AK076162.1 | 4 | 1646 | BAC36228.1 | |
| AK078708.1 | 4 | 1644 | ||
| AK078984.1 | 1 | 568 | ||
| AK150476.1 | 3 | 1485 | ||
| AK151053.1 | 4 | 1644 | BAE30070.1 | |
| AK159533.1 | 4 | 1612 | BAE35161.1 | |
| BC018559.1 | 4 | 1624 | AAH18559.1 |
More Information
Set Membership: |
|

Genomic Map