Assay Details
Target Gene Details
Entrez Gene ID: | 100040201 |
Gene Name: | predicted gene 10229 |
Gene Aliases: |
ENSMUSG00000068075 |
Location: |
Chr.16:89015277-89015846 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10229 | NM_001199334.2 | 1 | 474 | NP_001186263.1 |
| D86419.1 | 1 | 424 | BAA19680.1 |

Genomic Map