Assay Details
Target Gene Details
Entrez Gene ID: | 100040214 |
Gene Name: | predicted gene 10228 |
Gene Aliases: |
ENSMUSG00000068074 |
Location: |
Chr.16:89040923-89041472 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10228 | NM_001270487.1 | 1 | 364 | NP_001257416.1 |
| AK137757.1 | 1 | 365 | ||
| D86421.1 | 1 | 344 | BAA19682.1 |

Genomic Map