Assay Details
Target Gene Details
Entrez Gene ID: | 73916 |
Gene Name: | intraflagellar transport 57 |
Gene Aliases: |
4833420A15Rik, Esrrbl1, Hippi, MHS4R2 |
Location: |
Chr.16:49699294-49765126 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ift57 | NM_028680.3 | 11 | 1609 | NP_082956.2 |
| AK047217.1 | 11 | 1609 | BAC32995.1 | |
| BC138143.1 | 11 | 1583 | ||
| BC138144.1 | 11 | 1583 |
More Information
Set Membership: |
|

Genomic Map