Assay Details
Target Gene Details
Entrez Gene ID: | 16854 |
Gene Name: | lectin, galactose binding, soluble 3 |
Gene Aliases: |
GBP, L-34, Mac-2, gal3 |
Location: |
Chr.14:47373860-47386167 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgals3 | NM_001145953.1 | 3 | 198 | NP_001139425.1 |
| NM_010705.3 | 3 | 171 | NP_034835.1 | |
| AK008593.1 | BAE43211.1 | |||
| AK089249.1 | 3 | 115 | BAC40813.1 | |
| AK149783.1 | 3 | 145 | BAE29081.1 | |
| AK155244.1 | 3 | 114 | ||
| AK155484.1 | 3 | 113 | BAE33287.1 | |
| AK159536.1 | 3 | 139 | BAE35163.1 | |
| AK170803.1 | 3 | 228 | BAE42038.1 | |
| AK189964.1 | ||||
| BB853044.1 | 3 | 172 | ||
| BC132328.1 | 3 | 124 | ||
| BC138790.1 | 3 | 124 | ||
| BC145419.1 | 3 | 124 | ||
| J03723.1 | 1 | 71 | AAA37311.1 | |
| X16074.1 | 3 | 218 | CAA34206.1 | |
| X16834.1 | 3 | 117 | CAA34736.1 |
More Information
Set Membership: |
|

Genomic Map