Assay Details
Target Gene Details
Entrez Gene ID: | 16854 |
Gene Name: | lectin, galactose binding, soluble 3 |
Gene Aliases: |
GBP, L-34, Mac-2, gal3 |
Location: |
Chr.14:47373860-47386167 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgals3 | NM_001145953.1 | 6 | 865 | NP_001139425.1 |
| NM_010705.3 | 6 | 838 | NP_034835.1 | |
| AK008593.1 | 5 | 507 | BAE43211.1 | |
| AK089249.1 | 6 | 782 | BAC40813.1 | |
| AK149783.1 | 6 | 812 | BAE29081.1 | |
| AK155244.1 | 7 | 684 | ||
| AK155484.1 | 6 | 780 | BAE33287.1 | |
| AK159536.1 | 6 | 806 | BAE35163.1 | |
| AK170803.1 | 6 | 895 | BAE42038.1 | |
| BC132328.1 | 6 | 791 | ||
| BC138790.1 | 6 | 791 | ||
| BC145419.1 | 6 | 791 | ||
| J03723.1 | 4 | 738 | AAA37311.1 | |
| M33215.1 | 4 | 447 | AAA37314.1 | |
| X16074.1 | 6 | 885 | CAA34206.1 | |
| X16834.1 | 6 | 784 | CAA34736.1 |
More Information
Set Membership: |
|

Genomic Map