Assay Details
Target Gene Details
Entrez Gene ID: | 57755 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 7 |
Gene Aliases: |
4933424H20Rik, Dj5, mDj5 |
Location: |
Chr.15:81407088-81408273 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb7 | NM_021317.2 | 1 | 400 | NP_067292.2 |
| AB028855.1 | 1 | 400 | BAA88303.1 | |
| AK006201.1 | 1 | 426 | BAB24456.1 | |
| AK077186.1 | 1 | 429 | BAC36668.1 | |
| BC104396.1 | 1 | 347 | AAI04397.1 | |
| BC104397.1 | 1 | 347 | AAI04398.1 | |
| BC119478.2 | 1 | 347 |
Target Gene Details
Entrez Gene ID: | 321003 |
Gene Name: | X-prolyl aminopeptidase (aminopeptidase P) 3, putative |
Gene Aliases: |
APP3, E430012M05Rik |
Location: |
Chr.15:81400128-81457476 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xpnpep3 | NM_177310.2 | NP_796284.1 | ||
| AK042110.1 | ||||
| AK088324.1 | BAC40282.1 | |||
| AK158394.1 | ||||
| BC137569.1 | ||||
| BC144717.1 |
More Information
Set Membership: |
|

Genomic Map