Assay Details
Target Gene Details
Entrez Gene ID: | 72778 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 22 |
Gene Aliases: |
2810451A06Rik, AI506245 |
Location: |
Chr.15:99099134-99104737 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc22 | NM_176835.2 | 2 | 584 | NP_789805.1 |
| AK013323.1 | 2 | 608 | ||
| AK050294.1 | 2 | 562 | ||
| BC039633.1 | 2 | 584 | AAH39633.1 |
More Information
Set Membership: |
|

Genomic Map