Assay Details
Target Gene Details
Entrez Gene ID: | 432982 |
Gene Name: | predicted gene 5475 |
Gene Aliases: |
EG432982 |
Location: |
Chr.15:100423193-100428150 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5475 | NR_040351.1 | 1 | 868 | |
| AK163173.1 | 1 | 869 | BAE37220.1 | |
| BC147350.1 | 1 | 550 | ||
| BC147351.1 | 1 | 550 |
Target Gene Details
Entrez Gene ID: | 18174 |
Gene Name: | solute carrier family 11 (proton-coupled divalent metal ion transporters), member 2 |
Gene Aliases: |
DCT1, DMT1, Nramp2, mk, van |
Location: |
Chr.15:100387898-100424209 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc11a2 | AJ493663.1 | CAD38518.1 |
More Information
Set Membership: |
|

Genomic Map