Assay Details
Target Gene Details
Entrez Gene ID: | 432982 |
Gene Name: | predicted gene 5475 |
Gene Aliases: |
EG432982 |
Location: |
Chr.15:100423193-100428150 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5475 | NR_040351.1 | 3 | 1528 | |
| AK163173.1 | 3 | 1530 | BAE37220.1 | |
| BC147350.1 | 3 | 1210 | ||
| BC147351.1 | 3 | 1213 |

Genomic Map