Assay Details
Target Gene Details
Entrez Gene ID: | 671535 |
Gene Name: | poly (ADP-ribose) polymerase family, member 10 |
Gene Aliases: |
- |
Location: |
Chr.15:76232995-76243440 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp10 | NM_001163575.1 | 11 | 2998 | NP_001157047.1 |
| NM_001163576.1 | 10 | 3102 | NP_001157048.1 | |
| AK028373.1 | 11 | 2946 | BAC25913.1 | |
| AK035309.1 | 10 | 3122 | ||
| AK046755.1 | 10 | 3083 | BAC32856.1 | |
| AK146336.1 | 11 | 2998 | ||
| AK153205.1 | 10 | 2839 | BAE31805.1 | |
| AK166291.1 | 11 | 2949 | BAE38685.1 | |
| BC023056.1 | 7 | 1835 | AAH23056.1 | |
| BC024074.1 | 11 | 2955 | AAH24074.1 | |
| BC025608.1 | 7 | 1980 | AAH25608.1 | |
| BP768925.1 | 1 | 337 |

Genomic Map