Assay Details
Target Gene Details
Entrez Gene ID: | 66863 |
Gene Name: | leucine-zipper-like transcriptional regulator, 1 |
Gene Aliases: |
1200003E21Rik, AI591627, AW550890 |
Location: |
Chr.16:17508541-17526333 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lztr1 | NM_025808.3 | 21 | 2561 | NP_080084.2 |
| AK004561.2 | 21 | 2561 | BAB23373.2 | |
| AK005037.1 | 21 | 2560 | BAB23764.2 | |
| AK083411.1 | 21 | 2565 | BAC38905.1 | |
| AK088938.1 | 11 | 1666 | BAC40662.1 | |
| BC034400.1 | 21 | 2555 | AAH34400.1 |
More Information
Set Membership: |
|

Genomic Map