Assay Details
Target Gene Details
Entrez Gene ID: | 12374 |
Gene Name: | calcium-sensing receptor |
Gene Aliases: |
CaR, Gprc2a |
Location: |
Chr.16:36490585-36562141 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casr | NM_013803.2 | 4 | 1462 | NP_038831.2 |
| AF092941.1 | 1 | 249 | AAC62116.1 | |
| AF110178.1 | 4 | 1463 | AAD28371.1 | |
| AF110179.1 | 4 | 1463 | AAD28372.1 | |
| AF128842.1 | 3 | 1160 | AAD40638.1 | |
| AF159565.1 | 1 | 148 | AAF00193.1 | |
| AK029770.1 | 1 | 51 | BAC26608.1 |
More Information
Set Membership: |
|

Genomic Map