Assay Details
Target Gene Details
Entrez Gene ID: | 76872 |
Gene Name: | coiled-coil domain containing 116 |
Gene Aliases: |
4930432J16Rik |
Location: |
Chr.16:17139064-17147146 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc116 | NM_029779.2 | 4 | 2830 | NP_084055.2 |
| BC057359.1 | 4 | 3177 | AAH57359.1 |

Genomic Map