Assay Details
Target Gene Details
Entrez Gene ID: | 239839 |
Gene Name: | coiled-coil domain containing 14 |
Gene Aliases: |
G630039H03Rik |
Location: |
Chr.16:34690567-34725204 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc14 | NM_172824.3 | 8 | 1189 | NP_766412.2 |
| AK029725.1 | 7 | 2321 | ||
| AK090294.1 | 8 | 1096 | BAC41158.1 | |
| AK172014.1 | 2 | 2574 | ||
| BC031204.1 | 8 | 1082 | AAH31204.1 | |
| DV661299.1 |

Genomic Map