Assay Details
Target Gene Details
Entrez Gene ID: | 100504628 |
Gene Name: | predicted gene, 20319 |
Gene Aliases: |
- |
Location: |
Chr.16:26714682-26755502 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm20319 | NR_105063.1 | |||
| AA239010.1 |
Target Gene Details
Entrez Gene ID: | 16180 |
Gene Name: | interleukin 1 receptor accessory protein |
Gene Aliases: |
6430709H04Rik, AI255955, AV239853, IL-1RAcP |
Location: |
Chr.16:26581688-26730128 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1rap | NM_001159317.1 | 12 | 2166 | NP_001152789.1 |
| NM_001159318.1 | 11 | 2455 | NP_001152790.1 | |
| AK032620.1 | ||||
| AK033972.1 | 11 | 2404 | ||
| AK034269.1 | 12 | 2130 | ||
| AK083209.1 | 11 | 2452 | BAE43382.1 | |
| AK136782.1 | 11 | 2406 | BAE23128.1 |
More Information
Set Membership: |
|

Genomic Map