Assay Details
Target Gene Details
Entrez Gene ID: | 68479 |
Gene Name: | PHD finger protein 5A |
Gene Aliases: |
1110007B08Rik |
Location: |
Chr.15:81864516-81871892 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf5a | NM_026737.3 | 4 | 970 | NP_081013.1 |
| AK143590.1 | 4 | 971 | BAE25453.1 |

Genomic Map