Assay Details
Target Gene Details
Entrez Gene ID: | 71751 |
Gene Name: | mitogen-activated protein kinase kinase kinase 13 |
Gene Aliases: |
C130026N12Rik, LZK |
Location: |
Chr.16:21825171-21933439 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map3k13 | NM_172821.3 | 13 | 5395 | NP_766409.2 |
| AK047986.1 | 4 | 3469 | ||
| AK051378.1 | 4 | 3388 | BAC34618.1 | |
| AK054039.1 | 1 | 2231 | ||
| BB549158.2 | 1 | 97 |
More Information
Set Membership: |
|

Genomic Map