Assay Details
Target Gene Details
Entrez Gene ID: | 72083 |
Gene Name: | mitotic spindle organizing protein 2 |
Gene Aliases: |
2410018G20Rik, 2610001E06Rik, Fam128, Fam128b |
Location: |
Chr.16:15848441-15863364 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mzt2 | NM_029354.2 | 1 | 387 | NP_083630.1 |
| AK010544.1 | 1 | 388 | BAB27018.1 | |
| AK011255.1 | BAB27497.1 | |||
| BC027550.1 | 1 | 312 | AAH27550.1 |

Genomic Map