Assay Details
Target Gene Details
Entrez Gene ID: | 383103 |
Gene Name: | trans-golgi network vesicle protein 23A |
Gene Aliases: |
Fam18a |
Location: |
Chr.16:10420557-10447388 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tvp23a | NM_001013778.1 | 1 | 207 | NP_001013800.1 |
| BC068110.1 | 1 | 207 | AAH68110.1 |

Genomic Map