Assay Details
Target Gene Details
Entrez Gene ID: | 239283 |
Gene Name: | oxoglutarate (alpha-ketoglutarate) receptor 1 |
Gene Aliases: |
Cysltr3, Gm299, Gpr80, Gpr99, P2Y15 |
Location: |
Chr.14:120019585-120042435 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxgr1 | NM_001001490.3 | 4 | 2095 | NP_001001490.1 |
| AK142510.1 | 4 | 2096 | BAE25093.1 |
More Information
Set Membership: |
|

Genomic Map