Assay Details
Target Gene Details
Entrez Gene ID: | 67168 |
Gene Name: | lysophosphatidic acid receptor 6 |
Gene Aliases: |
2610302I02Rik, P2ry5, P2y5 |
Location: |
Chr.14:73237891-73240358 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lpar6 | NM_175116.4 | 1 | 415 | NP_780325.1 |
| AK008952.1 | 1 | 291 | ||
| AK011967.1 | 1 | 307 | ||
| AK032901.1 | 1 | 412 | BAC28077.1 | |
| AK169602.1 | 1 | 307 | BAE41252.1 | |
| BC047931.1 | 1 | 291 | AAH47931.1 | |
| BC069991.1 | 1 | 274 | AAH69991.1 | |
| BC145940.1 | 1 | 41 | ||
| BC145942.1 | 1 | 41 |
Target Gene Details
Entrez Gene ID: | 19645 |
Gene Name: | retinoblastoma 1 |
Gene Aliases: |
Rb, Rb-1, pRb |
Location: |
Chr.14:73194453-73325887 on Build GRCm38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rb1 | NM_009029.2 | NP_033055.2 | ||
| AK132875.1 | BAE21400.1 | |||
| AK141028.1 | BAE24548.1 | |||
| AK146467.1 | BAE27194.1 | |||
| BC096525.1 | AAH96525.1 | |||
| DQ400415.1 | ABD72475.1 | |||
| M26391.1 | AAA39964.1 |
More Information
Set Membership: |
|

Genomic Map