Assay Details
Target Gene Details
Entrez Gene ID: | 75409 |
Gene Name: | SLIT and NTRK-like family, member 5 |
Gene Aliases: |
2610019D03Rik |
Location: |
Chr.14:111675115-111683144 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slitrk5 | NM_198865.1 | 1 | 401 | NP_942565.1 |
| BC065107.1 | AAH65107.1 | |||
| BY274593.1 | 1 | 216 | ||
| BY728649.1 | 1 | 419 | ||
| CB526109.1 | 1 | 458 |

Genomic Map