Assay Details
Target Gene Details
Entrez Gene ID: | 218739 |
Gene Name: | sentan, cilia apical structure protein |
Gene Aliases: |
A430083B19Rik |
Location: |
Chr.14:13670876-13683148 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sntn | NM_177624.3 | 4 | 447 | NP_808292.1 |
| AK040281.1 | 4 | 448 | BAC30559.1 | |
| BC126945.1 | 4 | 448 |
More Information
Set Membership: |
|

Genomic Map