Assay Details
Target Gene Details
Entrez Gene ID: | 12297 |
Gene Name: | calcium channel, voltage-dependent, beta 3 subunit |
Gene Aliases: |
Beta3, CAB3, Ca(v)beta3, Cchb3 |
Location: |
Chr.15:98626873-98644536 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacnb3 | NM_001044741.2 | 1 | 145 | NP_001038206.1 |
| CD807951.1 | 1 | 253 |
More Information
Set Membership: |
|

Genomic Map