Assay Details
Target Gene Details
Entrez Gene ID: | 207393 |
Gene Name: | leucine rich repeat and fibronectin type III, extracellular 2 |
Gene Aliases: |
6330514E13, AW048948, BC094219, Lrrc62, Ppp1r29 |
Location: |
Chr.15:78667331-78718873 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elfn2 | NM_183141.2 | 2 | 3287 | NP_898964.2 |
| AK031970.1 | 1 | 2877 | BAC27630.1 | |
| AK173294.1 | 1 | 1753 | BAD32572.1 | |
| BC079588.1 | 1 | 2787 | AAH79588.1 | |
| BC094219.1 | 2 | 3287 | AAH94219.1 |
More Information
Set Membership: |
|

Genomic Map