Assay Details
Target Gene Details
Entrez Gene ID: | 68740 |
Gene Name: | keratin associated protein 22-2 |
Gene Aliases: |
1110032D16Rik |
Location: |
Chr.16:89010380-89010759 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap22-2 | NM_001191018.1 | 1 | 265 | NP_001177947.1 |
| AA763224.1 | 1 | 244 | ||
| AI646246.1 | 1 | 116 | ||
| AK004025.1 | 1 | 266 |

Genomic Map