Assay Details
Target Gene Details
Entrez Gene ID: | 239319 |
Gene Name: | caspase recruitment domain family, member 6 |
Gene Aliases: |
D730008L15 |
Location: |
Chr.15:5096920-5116483 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Card6 | NM_001163138.1 | 2 | 292 | NP_001156610.1 |
| AK052796.1 | 1 | 182 | BAC35150.1 |

Genomic Map