Assay Details
Target Gene Details
Entrez Gene ID: | 223527 |
Gene Name: | enhancer of yellow 2 homolog (Drosophila) |
Gene Aliases: |
1810057B09Rik, 6720481I12, DC6, Ey2 |
Location: |
Chr.15:44428081-44438256 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eny2 | NM_175009.3 | 5 | 1387 | NP_778174.1 |
| AK016562.1 | ||||
| AK032957.1 | 5 | 1387 | BAC28097.1 | |
| AK136334.1 | 1 | 474 | ||
| AK162511.1 | 5 | 1382 |
More Information
Set Membership: |
|

Genomic Map