Assay Details
Target Gene Details
Entrez Gene ID: | 252973 |
Gene Name: | grainyhead-like 2 (Drosophila) |
Gene Aliases: |
0610015A08Rik, BOM, Tcfcp2l3 |
Location: |
Chr.15:37233036-37363569 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grhl2 | NM_026496.4 | 4 | 761 | NP_080772.2 |
| AF411213.1 | 4 | 494 | AAM22619.1 | |
| AK002633.1 | 4 | 762 | BAB22247.1 | |
| BC055035.1 | 4 | 550 | AAH55035.1 | |
| BY703140.1 | 4 | 761 |
More Information
Set Membership: |
|

Genomic Map