Assay Details
Target Gene Details
Entrez Gene ID: | 29857 |
Gene Name: | mitogen-activated protein kinase 12 |
Gene Aliases: |
AW123708, Erk6, P38gamma, Prkm12, Sapk3 |
Location: |
Chr.15:89130584-89141593 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk12 | NM_013871.3 | 12 | 1473 | NP_038899.1 |
| AA855742.1 | 2 | 262 | ||
| AI839277.1 | ||||
| AK011286.1 | 11 | 1513 | ||
| AK206111.1 | ||||
| AK207281.1 | ||||
| BC002134.1 | 2 | 1014 | AAH02134.1 | |
| BC021640.1 | 12 | 1391 | AAH21640.1 | |
| Y13439.1 | 12 | 1409 | CAA73850.1 |
More Information
Set Membership: |
|

Genomic Map