Assay Details
Target Gene Details
Entrez Gene ID: | 106068 |
Gene Name: | solute carrier family 45, member 4 |
Gene Aliases: |
9330175B01Rik, AW552276 |
Location: |
Chr.15:73577418-73645757 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc45a4 | NM_001033219.3 | 8 | 2997 | NP_001028391.2 |
| NM_001168255.1 | 7 | 2075 | NP_001161727.1 | |
| AK133269.1 | 8 | 2998 | BAE21586.1 | |
| AK170738.1 | 7 | 2080 | BAE41991.1 | |
| AK173104.1 | 4 | 1254 | BAD32382.1 | |
| BC056501.1 | 8 | 2594 | ||
| BC058722.1 | 7 | 1993 | AAH58722.1 |
More Information
Set Membership: |
|

Genomic Map