Assay Details
Target Gene Details
Entrez Gene ID: | 57277 |
Gene Name: | secreted Ly6/Plaur domain containing 1 |
Gene Aliases: |
1110021N19Rik, AI415082, ARS, ArsB, Slurp-1 |
Location: |
Chr.15:74726644-74728026 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slurp1 | NM_020519.1 | 3 | 479 | NP_065265.1 |
| AK003904.1 | BAB23068.1 |

Genomic Map