Assay Details
Target Gene Details
Entrez Gene ID: | 210544 |
Gene Name: | TBC1 domain family, member 31 |
Gene Aliases: |
4B3, D330013L20Rik, Gm85, Wdr67 |
Location: |
Chr.15:57911684-57970068 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d31 | NM_001081396.2 | 21 | 3029 | NP_001074865.2 |
| BC050039.1 | 21 | 2975 | AAH50039.1 | |
| BC066830.1 | 21 | 2961 | AAH66830.1 |

Genomic Map