Assay Details
Target Gene Details
Entrez Gene ID: | 22402 |
Gene Name: | WNT1 inducible signaling pathway protein 1 |
Gene Aliases: |
AW146261, CCN4, Elm1 |
Location: |
Chr.15:66891325-66923205 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wisp1 | NM_018865.2 | 5 | 1652 | NP_061353.1 |
| AB004873.1 | 5 | 1652 | BAA24949.1 | |
| AF100777.1 | 5 | 1557 | AAC96319.1 | |
| AK013300.1 | 5 | 1676 | ||
| AK148457.1 | 5 | 1674 | BAE28565.1 | |
| BC048791.1 | 5 | 1466 | AAH48791.1 | |
| BC052677.1 | 6 | 1626 | AAH52677.1 |
More Information
Set Membership: |
|

Genomic Map