Assay Details
Target Gene Details
Entrez Gene ID: | 12519 |
Gene Name: | CD80 antigen |
Gene Aliases: |
B71, Cd28l, Ly-53, Ly53, MIC17, TSA1 |
Location: |
Chr.16:38455568-38498546 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd80 | NM_009855.2 | 1 | 301 | NP_033985.3 |
| AF065893.1 | 1 | 110 | AAD25876.1 | |
| AF065894.1 | 1 | 110 | AAD25877.1 | |
| AF065895.1 | 1 | 110 | AAD25878.1 | |
| AF065896.1 | 1 | 110 | AAD25879.1 | |
| AJ278965.1 | 1 | 53 | CAC34306.1 | |
| AK154329.1 | 2 | 410 | BAE32518.1 | |
| AY278186.1 | 1 | 101 | AAP57303.1 | |
| BC131959.1 | 1 | 121 | ||
| BC145843.1 | 1 | 121 | ||
| D16220.1 | 1 | 116 | BAA03748.1 | |
| X60958.1 | 1 | 301 | CAA43291.1 |
More Information
Set Membership: |
|

Genomic Map