Assay Details
Target Gene Details
Entrez Gene ID: | 72003 |
Gene Name: | synaptoporin |
Gene Aliases: |
1500003F20Rik, SPO |
Location: |
Chr.14:13284780-13615469 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synpr | NM_001163032.1 | 1 | 121 | NP_001156504.1 |
| NM_028052.4 | NP_082328.3 | |||
| AK005132.1 | BAB23831.1 | |||
| AK032442.1 | BAC27871.1 | |||
| AK033999.1 | 1 | 95 | ||
| AK045705.1 | ||||
| AK049661.1 | BAC33864.1 | |||
| AK078223.1 | BAC37181.1 | |||
| BC026512.1 | AAH26512.1 | |||
| CD354682.1 | 1 | 121 |
More Information
Set Membership: |
|

Genomic Map