Assay Details
Target Gene Details
Entrez Gene ID: | 75729 |
Gene Name: | family with sequence similarity 227, member A |
Gene Aliases: |
4933432B09Rik |
Location: |
Chr.15:79609576-79658970 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam227a | NM_029407.1 | 18 | 2229 | NP_083683.1 |
| AK017017.1 | 18 | 2068 | BAB30551.1 | |
| BC118956.1 | 15 | 1731 |

Genomic Map