Assay Details
Target Gene Details
Entrez Gene ID: | 69659 |
Gene Name: | RIKEN cDNA 2310069G16 gene |
Gene Aliases: |
- |
Location: |
Chr.15:44787763-44805829 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2310069G16Rik | NR_040309.1 | 1 | 58 | |
| AK010129.1 | 1 | 40 | ||
| CJ061573.1 | 1 | 59 |
Target Gene Details
Entrez Gene ID: | 319613 |
Gene Name: | syntabulin (syntaxin-interacting) |
Gene Aliases: |
Golsyn |
Location: |
Chr.15:44671856-44788063 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sybu | NM_001032727.1 | 1 | 244 | NP_001027899.1 |
| NM_176998.4 | 1 | 244 | NP_795972.2 | |
| AB232447.1 | 1 | 244 | BAE19961.1 | |
| AB232448.1 | 1 | 244 | BAE19962.1 | |
| AK043739.1 | 1 | 244 | BAC31638.1 | |
| AK047449.1 | 1 | 234 | ||
| AK049811.1 | 1 | 178 | BAC33929.1 |
More Information
Set Membership: |
|

Genomic Map