Assay Details
Target Gene Details
Entrez Gene ID: | 207911 |
Gene Name: | melanin-concentrating hormone receptor 1 |
Gene Aliases: |
AW049955, Gpr24, Mch1r |
Location: |
Chr.15:81235499-81238964 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mchr1 | NM_145132.2 | 2 | 1165 | NP_660114.1 |
| AF498247.1 | 2 | 890 | AAM22964.1 | |
| AF498248.1 | 2 | 890 | AAM22965.1 | |
| AK134867.1 | 2 | 1048 | BAE22319.1 | |
| BC128286.1 | 2 | 890 |
More Information
Set Membership: |
|

Genomic Map