Assay Details
Target Gene Details
Entrez Gene ID: | 20649 |
Gene Name: | syntrophin, basic 1 |
Gene Aliases: |
- |
Location: |
Chr.15:55638505-55907305 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sntb1 | NM_016667.3 | 7 | 1923 | NP_057876.1 |
| U89997.1 | 7 | 1928 | AAB66697.1 |

Genomic Map