Assay Details
Target Gene Details
Entrez Gene ID: | 14375 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 6 |
Gene Aliases: |
70kDa, G22p1, Ku70 |
Location: |
Chr.15:81987826-82040085 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrcc6 | NM_010247.2 | 13 | 1806 | NP_034377.2 |
| AB010282.1 | 13 | 1747 | BAA28874.1 | |
| AF483500.1 | 12 | 1686 | AAL90774.1 | |
| AF483501.1 | 12 | 1686 | AAL90775.1 | |
| AK088530.1 | 7 | 756 | ||
| AK143570.1 | 13 | 1781 | BAE25441.1 | |
| AK146392.1 | 13 | 1725 | BAE27135.1 | |
| AK146394.1 | 13 | 1725 | BAE27137.1 | |
| AK146396.1 | 13 | 1773 | BAE27139.1 | |
| BC031422.1 | 13 | 1755 | AAH31422.1 | |
| M38700.1 | 12 | 1648 | AAA39396.1 |
More Information
Set Membership: |
|

Genomic Map