Assay Details
Target Gene Details
Entrez Gene ID: | 17225 |
Gene Name: | mast cell protease 2 |
Gene Aliases: |
MMCP-2, Mcp-2 |
Location: |
Chr.14:56021584-56044635 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcpt2 | NM_008571.1 | 5 | 859 | NP_032597.1 |
| AK133521.1 | 5 | 860 | BAE21702.1 | |
| AY007570.1 | 5 | 859 | AAG24504.1 | |
| BC119248.1 | ||||
| BC119250.1 | ||||
| J05177.1 | 5 | 841 | AAA39972.1 |
More Information
Set Membership: |
|

Genomic Map