Assay Details
Target Gene Details
Entrez Gene ID: | 319613 |
Gene Name: | syntabulin (syntaxin-interacting) |
Gene Aliases: |
Golsyn |
Location: |
Chr.15:44671856-44788063 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sybu | NM_001032727.1 | 6 | 2332 | NP_001027899.1 |
| NM_001285840.1 | 7 | 2438 | NP_001272769.1 | |
| NM_001285841.1 | 4 | 1679 | NP_001272770.1 | |
| NM_001285842.1 | 5 | 1823 | NP_001272771.1 | |
| NM_001285843.1 | 6 | 1890 | NP_001272772.1 | |
| NM_001285844.1 | 5 | 1782 | NP_001272773.1 | |
| NM_176998.4 | 8 | 2639 | NP_795972.2 | |
| NM_178765.3 | 8 | 2536 | NP_848880.2 | |
| AB232447.1 | 8 | 2639 | BAE19961.1 | |
| AB232448.1 | 6 | 2332 | BAE19962.1 | |
| AB232449.1 | 8 | 2536 | BAE19963.1 | |
| AK029072.1 | 7 | 2135 | BAC26277.1 | |
| AK030482.1 | 8 | 2268 | BAC26984.1 | |
| AK030720.1 | 9 | 2268 | BAC27098.1 | |
| AK163432.1 | 8 | 2301 | BAE37345.1 | |
| AK220268.1 | 7 | 2438 | BAD90193.1 | |
| BC048945.1 | 6 | 2064 | AAH48945.1 | |
| BC065045.1 | 8 | 2533 | AAH65045.1 |
More Information
Set Membership: |
|

Genomic Map