Assay Details
Target Gene Details
Entrez Gene ID: | 104445 |
Gene Name: | CDC42 effector protein (Rho GTPase binding) 1 |
Gene Aliases: |
1810058K22Rik, AA980734, Borg5, CEP1, MSE55 |
Location: |
Chr.15:78842647-78850902 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42ep1 | NM_027219.3 | 2 | 437 | NP_081495.1 |
| AK007896.1 | 2 | 438 | BAB25335.1 | |
| BC016250.1 | 2 | 425 | AAH16250.1 | |
| BC083130.1 | 2 | 430 | AAH83130.1 |

Genomic Map