Assay Details
Target Gene Details
Entrez Gene ID: | 66270 |
Gene Name: | family with sequence similarity 134, member B |
Gene Aliases: |
1810015C04Rik, AU015349 |
Location: |
Chr.15:25843298-25973696 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam134b | NM_001034851.2 | 1 | 197 | NP_001030023.1 |
| BY779369.1 | ||||
| CB724233.1 | 1 | 315 |

Genomic Map