Assay Details
Target Gene Details
Entrez Gene ID: | 15202 |
Gene Name: | hematopoietic cell transcript 1 |
Gene Aliases: |
1700057K19Rik, HemT |
Location: |
Chr.15:74818880-74846164 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hemt1 | NM_010416.2 | 4 | 511 | NP_034546.2 |
| AJ011022.1 | 2 | 434 | CAB39201.1 | |
| AJ011023.1 | 2 | 396 | CAB39202.1 | |
| AK005659.1 | 4 | 516 | BAB24171.1 | |
| AK005823.1 | 3 | 284 | ||
| AK006826.1 | ||||
| BC119337.1 | 2 | 427 | ||
| DQ701705.1 |

Genomic Map